Dibenzothiophen
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
Dibenzo[b,d]thiophen
, Diphenylensulfid
ACTIAS32977G100EA
0
CHF
ACTIAS32977G100
Dibenzothiophen
Dibenzothiophen
Formel:
C₁₂H₈S MW: 184,26 g/mol Siedepunkt: 332 °C (1013 hPa) Melting Pt: 95…98 °C Dichte: 1,1295 g/cm³ (125 °C) Flash-Pt: 170 °C |
MDL Number:
MFCD00004969 CAS-Nummer: 132-65-0 EINECS: 205-072-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...