7-Amino-1H-indazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS21073G5EA 0 CHF
ACTIAS21073G5 ACTIAS21073G1
7-Amino-1H-indazole
7-Amino-1H-indazole
Formula: C₇H₇N₃
MW: 133.15 g/mol
MDL Number: MFCD00022790
CAS Number: 21443-96-9
EINECS: 244-391-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR