6-Nitro-1H-indazole

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 6-NINA6-Nitroindazole6-Nitro-1H-indazole

ACTIAS10189G5EA 0 CHF
ACTIAS10189G1 ACTIAS10189MG500 ACTIAS10189G5
6-Nitro-1H-indazole
6-Nitro-1H-indazole
Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
Melting Pt: 181 °C
MDL Number: MFCD00005695
CAS Number: 7597-18-4
EINECS: 231-500-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR