6-Nitrobenzothiazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31383G5EA 0 CHF
ACTIAS31383G5 ACTIAS31383G10
6-Nitrobenzothiazole
6-Nitrobenzothiazole
Formula: C₇H₄N₂O₂S
MW: 180.19 g/mol
Melting Pt: 178 °C
MDL Number: MFCD00014569
CAS Number: 2942-06-5
EINECS: 220-933-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR