Indole-6-carbaldehyd

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1H-Indole-6-carbaldehydeIndole-6-carboxaldehyde

ACTIAS50082G1EA 0 CHF
ACTIAS50082G1 ACTIAS50082G25 ACTIAS50082G5
Indole-6-carbaldehyd
Indole-6-carbaldehyd
Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD02179596
CAS Number: 1196-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR