6-(Trifluoromethyl)indole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS37553G5EA 0 CHF
ACTIAS37553G10 ACTIAS37553G5
6-(Trifluoromethyl)indole
6-(Trifluoromethyl)indole
Formula: C₉H₆F₃N
MW: 185.15 g/mol
Melting Pt: 109…110 °C
MDL Number: MFCD00272316
CAS Number: 13544-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR