5-Phenyl-1H-tetrazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS33064G10EA 0 CHF
ACTIAS33064G10
5-Phenyl-1H-tetrazole
5-Phenyl-1H-tetrazole
Formula: C₇H₆N₄
MW: 146.15 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 222 °C
MDL Number: MFCD00022388
CAS Number: 18039-42-4
EINECS: 241-950-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR