5-Methylphenazinium methyl sulphate
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Danger
|
Synonyms:
Phenazine methosulphate
, PMS
ACTIAS23185G25EA
0
CHF
ACTIAS23185G25
ACTIAS23185G5
5-Methylphenazinium methyl sulphate
5-Methylphenazinium methyl sulphate
Formula:
C₁₄H₁₄N₂O₄S MW: 306.34 g/mol Melting Pt: 198 °C Storage Temperature: Freezer |
MDL Number:
MFCD00011923 CAS Number: 299-11-6 EINECS: 206-072-1 Merck Index: 12,06185 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...