5-Methyl-3-isoxazolcarbonsäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS8473G5EA
0
CHF
ACTIAS8473G1
ACTIAS8473G5
5-Methyl-3-isoxazolcarbonsäure
5-Methylisoxazole-3-carboxylic acid
Formel:
C₅H₅NO₃ MW: 127,1 g/mol |
MDL Number:
MFCD01318162 CAS-Nummer: 3405-77-4 EINECS: 222-289-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...