5-Bromo-2-chloropyrimidine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS51681G5EA 0 CHF
ACTIAS51681G5 ACTIAS51681G25
5-Bromo-2-chloropyrimidine
5-Bromo-2-chloropyrimidine
Formula: Câ‚„Hâ‚‚BrClNâ‚‚
MW: 193.43 g/mol
MDL Number: MFCD00483232
CAS Number: 32779-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR