5-Bromo-2,4-dichloropyrimidine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS51683G25EA 0 CHF
ACTIAS51683G5 ACTIAS51683G25
5-Bromo-2,4-dichloropyrimidine
5-Bromo-2,4-dichloropyrimidine
Formula: C₄HBrCl₂N₂
MW: 227.87 g/mol
Boiling Pt: 288 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1.781 g/cm³ (20 °C)
MDL Number: MFCD00127818
CAS Number: 36082-50-5
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR