5-(4-Chlorophenyl)-1H-tetrazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS23656G25EA 261 CHF
ACTIAS23656G25
5-(4-Chlorophenyl)-1H-tetrazole
5-(4-Chlorophenyl)-1H-tetrazole
Formula: C₇H₅ClN₄
MW: 180.6 g/mol
MDL Number: MFCD00040952
CAS Number: 16687-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR