4-Nitro-1H-indazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS21072G5EA 55 CHF
ACTIAS21072G5 ACTIAS21072G1
4-Nitro-1H-indazole
4-Nitro-1H-indazole
Formula: C₇H₅N₃O₂
MDL Number: MFCD00022784
CAS Number: 2942-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR