4-Methylthiazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS33460G100EA 0 CHF
ACTIAS33460G10 ACTIAS33460G100
4-Methylthiazole
4-Methylthiazole
Formula: Câ‚„Hâ‚…NS
MW: 99.16 g/mol
Boiling Pt: 133 °C (1013 hPa)
Density: 1,1187 g/cm³ (20 °C)
MDL Number: MFCD00005340
CAS Number: 693-95-8
EINECS: 211-764-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR