Indole-4-carbaldehyd

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Indole-4-carboxaldehyde1H-Indole-4-carbaldehyde

ACTIAS7164G5EA 0 CHF
ACTIAS7164G5 ACTIAS7164G100
Indole-4-carbaldehyd
Indole-4-carbaldehyd
Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD01632221
CAS Number: 1074-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR