4-Amino-2-chloropyrimidine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Chloropyrimidin-4-ylamine2-Chloro-4-pyrimidinamine2-Chloropyrimidin-4-amine2-Chlor-4-pyrimidinamin

ACTIAS30438G5EA 0 CHF
ACTIAS30438G1 ACTIAS30438G5
4-Amino-2-chloropyrimidine
4-Amino-2-chloropyrimidine
Formula: C₄H₄ClN₃
MW: 129.55 g/mol
MDL Number: MFCD00194051
CAS Number: 7461-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR