4-Amino-2,6-dichloropyrimidine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS1406G100EA 0 CHF
ACTIAS1406G25 ACTIAS1406G100
4-Amino-2,6-dichloropyrimidine
4-Amino-2,6-dichloropyrimidine
Formula: C₄H₃Cl₂N₃
MW: 163.99 g/mol
MDL Number: MFCD00038015
CAS Number: 10132-07-7
EINECS: 233-369-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR