4,6-Dihydroxypyrimidine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 6-Hydroxy-1H-pyrimidin-4-one

ACTIAS20135G100EA 94 CHF
ACTIAS20135G100 ACTIAS20135G500
4,6-Dihydroxypyrimidine
4,6-Dihydroxypyrimidine
Formula: Câ‚„Hâ‚„Nâ‚‚Oâ‚‚
MW: 112.09 g/mol
Melting Pt: > 300 °C
MDL Number: MFCD00016733
CAS Number: 1193-24-4
EINECS: 214-772-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR