4,6-Dichloropyrimidine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS50160G100EA 0 CHF
ACTIAS50160G100
4,6-Dichloropyrimidine
4,6-Dichloropyrimidine
Formula: Câ‚„Hâ‚‚Clâ‚‚Nâ‚‚
MW: 148.98 g/mol
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: 64…66 °C
Density: 1.734 g/cm³ (20 °C)
MDL Number: MFCD00006109
CAS Number: 1193-21-1
EINECS: 214-770-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR