4,5-Dimethylthiazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS24097G100EA 142 CHF
ACTIAS24097G100
4,5-Dimethylthiazole
4,5-Dimethylthiazole
Formula: C₅H₇NS
MW: 113.18 g/mol
Boiling Pt: 149…153 °C (1013 hPa)
Density: 1,0828 g/cm³ (20 °C)
MDL Number: MFCD00005336
CAS Number: 3581-91-7
EINECS: 222-703-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR