Indole-3-carbaldehyd

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1H-Indole-3-carbaldehydeIndole-3-carboxaldehyde

ACTIAS7109G100EA 0 CHF
ACTIAS7109G100 ACTIAS7109KG1
Indole-3-carbaldehyd
Indole-3-carbaldehyd
Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 192…195 °C
Density: 1,34 g/cm³ (20 °C)
MDL Number: MFCD00005622
CAS Number: 487-89-8
EINECS: 207-665-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR