3-Acetylindol

Lieferant: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-Indolyl methyl ketoneIndol-3-ylmethylketon3-Indolylmethylketon1-(3-Indolyl)-1-ethanon

ACTIAS21126G25EA 50 CHF
ACTIAS21126G25 ACTIAS21126G100
3-Acetylindol
3-Acetylindol
Formel: C₁₀H₉NO
MW: 159,19 g/mol
Siedepunkt: 144…145 °C (10 mmHg)
Melting Pt: 186…189 °C
MDL Number: MFCD00005626
CAS-Nummer: 703-80-0
EINECS: 211-875-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR