3-Chloro-2-thenoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-Chloro-2-thiophenecarboxylic acid3-Chlorothiophene-2-carboxylic acid

ACTIAS22593G25EA 199 CHF
ACTIAS22593G25
3-Chloro-2-thenoic acid
3-Chloro-2-thenoic acid
Formula: C₅H₃ClO₂S
MW: 162.6 g/mol
Melting Pt: 187…188 °C
MDL Number: MFCD00043888
CAS Number: 59337-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR