3-Bromo-2-thenoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31855G5EA 0 CHF
ACTIAS31855G1 ACTIAS31855G5
3-Bromo-2-thenoic acid
3-Bromo-2-thenoic acid
Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
MDL Number: MFCD00052291
CAS Number: 7311-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR