5-Amino-2-methoxypyridine
Supplier: ACTIVATE SCIENTIFIC GMBH
ACTIAS30951G5EA
0
CHF
ACTIAS30951G5
ACTIAS30951G10
5-Amino-2-methoxypyridine
5-Amino-2-methoxypyridine
Formula:
C₆H₈N₂O MW: 124.14 g/mol Boiling Pt: 265 °C (1013 hPa) Melting Pt: 28…30 °C |
MDL Number:
MFCD00006264 CAS Number: 6628-77-9 EINECS: 229-612-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...