5-Amino-2-(trifluormethyl)pyridin
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
2-(Trifluormethyl)-5-pyridylamin
, 2-(Trifluormethyl)pyridin-5-amin
ACTIAS50419G5EA
0
CHF
ACTIAS50419G5
ACTIAS50419G1
ACTIAS50419G25
5-Amino-2-(trifluormethyl)pyridin
5-Amino-2-(trifluormethyl)pyridin
Formel:
C₆H₅F₃N₂ MW: 162,11 g/mol |
MDL Number:
MFCD01862656 CAS-Nummer: 106877-33-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...