3,5-Dibromobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40282G5EA 0 CHF
ACTIAS40282G10 ACTIAS40282G5
3,5-Dibromobenzaldehyde
3,5-Dibromobenzaldehyde
Formula: C₇H₄Br₂O
MW: 263.92 g/mol
MDL Number: MFCD00156887
CAS Number: 56990-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR