3-(4-Aminophenyl)propionsäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
3-(4-Aminophenyl)propansäure
ACTIAS35050G5EA
0
CHF
ACTIAS35050G5
ACTIAS35050G10
ACTIAS35050G1
3-(4-Aminophenyl)propionsäure
3-(4-Aminophenyl)propionsäure
Formel:
C₉H₁₁NO₂ MW: 165,19 g/mol |
MDL Number:
MFCD00017118 CAS-Nummer: 2393-17-1 EINECS: 219-245-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...