3-(3-Bromphenyl)propionsäure

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS40264G5EA 0 CHF
ACTIAS40264G25 ACTIAS40264G5
3-(3-Bromphenyl)propionsäure
3-(3-Bromphenyl)propionsäure
Formel: C₉H₉BrO₂
MW: 229,07 g/mol
MDL Number: MFCD01310792
CAS-Nummer: 42287-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR