2-Naphthalenesulphonyl chloride

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: Naphthalene-2-sulphonyl chlorideβ-Naphthalenesulphonyl chloride

ACTIAS20986G100EA 0 CHF
ACTIAS20986G100
2-Naphthalenesulphonyl chloride
2-Naphthalenesulphonyl chloride
Formula: C₁₀H₇ClO₂S
MW: 226.68 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 71…72 °C
MDL Number: MFCD00004087
CAS Number: 93-11-8
EINECS: 202-219-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR