2-Methyl-5-nitrobenzoxazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS25456G5EA 225 CHF
ACTIAS25456G5
2-Methyl-5-nitrobenzoxazole
2-Methyl-5-nitrobenzoxazole
Formula: C₈H₆N₂O₃
MW: 178.15 g/mol
MDL Number: MFCD00466748
CAS Number: 32046-51-8
EINECS: 250-904-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR