6-Fluor-o-anisaldehyd

Lieferant: ACTIVATE SCIENTIFIC GMBH

Synonyms: 6-Fluor-2-anisaldehyd2-Fluor-6-methoxybenzaldehyd

ACTIAS35186G1EA 0 CHF
ACTIAS35186G1 ACTIAS35186G10 ACTIAS35186G5
6-Fluor-o-anisaldehyd
6-Fluor-o-anisaldehyd
Formel: C₈H₇FO₂
MW: 154,14 g/mol
MDL Number: MFCD01090998
CAS-Nummer: 146137-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR