2-Chlorothiazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS50261G5EA 0 CHF
ACTIAS50261G5 ACTIAS50261G1
2-Chlorothiazole
2-Chlorothiazole
Formula: C₃H₂ClNS
MW: 119.57 g/mol
MDL Number: MFCD00210701
CAS Number: 3034-52-4
EINECS: 221-228-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR