2-Chlorobenzothiazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40222G100EA 0 CHF
ACTIAS40222G100
2-Chlorobenzothiazole
2-Chlorobenzothiazole
Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: 21…24 °C
Density: 1,37 g/cm³ (20 °C)
MDL Number: MFCD00005776
CAS Number: 615-20-3
EINECS: 210-415-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR