2-Chloroisonicotinonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Chloropyridine-4-carbonitrile2-Chloro-4-cyanopyridine

ACTIAS40205G100EA 0 CHF
ACTIAS40205G100
2-Chloroisonicotinonitrile
2-Chloroisonicotinonitrile
Formula: C₆H₃ClN₂
MW: 138.56 g/mol
MDL Number: MFCD00174318
CAS Number: 33252-30-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR