2-Bromo-5-nitrothiazole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS50072G5EA 0 CHF
ACTIAS50072G25 ACTIAS50072G5
2-Bromo-5-nitrothiazole
2-Bromo-5-nitrothiazole
Formula: C₃HBrN₂O₂S
MW: 209.02 g/mol
Melting Pt: 93 °C
Density: 2,295 g/cm³ (20 °C)
MDL Number: MFCD00005317
CAS Number: 3034-48-8
EINECS: 221-226-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR