2-Bromo-5-(trifluoromethyl)pyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS4052G5EA 0 CHF
ACTIAS4052G25 ACTIAS4052G5
2-Bromo-5-(trifluoromethyl)pyridine
2-Bromo-5-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
MW: 226 g/mol
MDL Number: MFCD00153086
CAS Number: 50488-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR