2-Bromoisonicotinonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS33596G1EA 0 CHF
ACTIAS33596G10 ACTIAS33596G1 ACTIAS33596G5
2-Bromoisonicotinonitrile
2-Bromoisonicotinonitrile
Formula: C₆H₃BrN₂
MW: 183.01 g/mol
MDL Number: MFCD07367879
CAS Number: 10386-27-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR