2-Bromo-3-(trifluoromethyl)pyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS7124G5EA 0 CHF
ACTIAS7124G25 ACTIAS7124G5
2-Bromo-3-(trifluoromethyl)pyridine
2-Bromo-3-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
MW: 226 g/mol
MDL Number: MFCD00153084
CAS Number: 175205-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR