2-Amino-5-phenylpyridine
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
5-Phenyl-2-pyridylamine
, 5-Phenylpyridin-2-amine
ACTIAS35338G10EA
0
CHF
ACTIAS35338G10
ACTIAS35338G1
ACTIAS35338G5
2-Amino-5-phenylpyridine
2-Amino-5-phenylpyridine
Formula:
C₁₁H₁₀N₂ MW: 170.21 g/mol |
MDL Number:
MFCD01692452 CAS Number: 33421-40-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...