2-Amino-5-(trifluoromethyl)pyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-(Trifluoromethyl)-2-pyridylamine5-(Trifluoromethyl)pyridin-2-amine

ACTIAS4020G100EA 0 CHF
ACTIAS4020G25 ACTIAS4020G100
2-Amino-5-(trifluoromethyl)pyridine
2-Amino-5-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
MDL Number: MFCD00042164
CAS Number: 74784-70-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR