2-Amino-4,6-dimethoxypyrimidine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS1408G100EA 0 CHF
ACTIAS1408G100 ACTIAS1408G25
2-Amino-4,6-dimethoxypyrimidine
2-Amino-4,6-dimethoxypyrimidine
Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 97 °C
Density: 1,429 g/cm³ (–153 °C)
MDL Number: MFCD00038832
CAS Number: 36315-01-2
EINECS: 252-969-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR