2,7-Dibromonaphthalene

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31988G5EA 0 CHF
ACTIAS31988G1 ACTIAS31988G5
2,7-Dibromonaphthalene
2,7-Dibromonaphthalene
Formula: C₁₀H₆Br₂
MW: 285.97 g/mol
MDL Number: MFCD00238580
CAS Number: 58556-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR