5-Ethyluracil

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Ethyluracil

ACTIAS20599G25EA 45 CHF
ACTIAS20599G25 ACTIAS20599G5 ACTIAS20599G100
5-Ethyluracil
5-Ethyluracil
Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD00079187
CAS Number: 4212-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR