2-(Trifluoromethyl)pyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31485G5EA 0 CHF
ACTIAS31485G1 ACTIAS31485G5
2-(Trifluoromethyl)pyridine
2-(Trifluoromethyl)pyridine
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 138…139 °C (1013 hPa)
MDL Number: MFCD02183562
CAS Number: 368-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR