5-Fluoro-o-toluonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS24178G100EA 289 CHF
ACTIAS24178G100
5-Fluoro-o-toluonitrile
5-Fluoro-o-toluonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 42…46 °C
MDL Number: MFCD00042295
CAS Number: 77532-79-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR