2-Cyanphenylisocyanat
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS36425G5EA
0
CHF
ACTIAS36425G5
ACTIAS36425MG250
ACTIAS36425G1
2-Cyanphenylisocyanat
2-Cyanphenylisocyanat
Formel:
C₈H₄N₂O MW: 144,13 g/mol |
CAS-Nummer:
42066-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...