2,5-Bis(trifluoromethyl)benzonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS25271G25EA 0 CHF
ACTIAS25271G25
2,5-Bis(trifluoromethyl)benzonitrile
2,5-Bis(trifluoromethyl)benzonitrile
Formula: C₉H₃F₆N
MW: 239.12 g/mol
MDL Number: MFCD03094423
CAS Number: 51012-27-2
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR