5-Chlorovaleronitrile

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 5-Chloropentanenitrile

ACTIAS21628G5EA 28 CHF
ACTIAS21628G5 ACTIAS21628G100 ACTIAS21628G25 ACTIAS21628G500
5-Chlorovaleronitrile
5-Chlorovaleronitrile
Formula: C₅H₈ClN
MW: 117.58 g/mol
Boiling Pt: 92 °C (1013 hPa)
Density: 1.447 g/cm³ (20 °C)
MDL Number: MFCD00001977
CAS Number: 6280-87-1
EINECS: 228-481-4
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR