3-Chloro-4-fluorobenzotrifluoride

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS24814G25EA 0 CHF
ACTIAS24814G25 ACTIAS24814G100
3-Chloro-4-fluorobenzotrifluoride
3-Chloro-4-fluorobenzotrifluoride
Formula: C₇H₃ClF₄
MW: 198.55 g/mol
MDL Number: MFCD00042207
CAS Number: 78068-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR